Tisk…

2-Bromo-4-fluoro-N-(2-fluorobenzyl)aniline, 97%

H57029.03EA 11400 CZK
ALFAH57029.03 ALFAH57029.MD
2-Bromo-4-fluoro-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1038976-20-3
Molecular Formula: C13H10BrF2N
Formula Weight: 298.13
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR