Tisk…

2,3-Dichloro-N-(4-fluorobenzyl)aniline, 97%

H57877.MDEA 2450 CZK
ALFAH57877.MD ALFAH57877.03
2,3-Dichloro-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1036453-69-6
Molecular Formula: C13H10Cl2FN
Formula Weight: 270.13
MDL No.: MFCD11117043
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR