Tisk…

2,4-Dichloro-N-(3-fluorobenzyl)aniline, 97%

H57748.MDEA 610 CZK
ALFAH57748.MD ALFAH57748.03
2,4-Dichloro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1036576-63-2
Molecular Formula: C13H10Cl2FN
Formula Weight: 270.13
MDL No.: MFCD11118672
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR