Tisk…

3-Chloro-N-(3-fluorobenzyl)-4-methoxyaniline, 97%

H57358.MDEA 1020 CZK
ALFAH57358.MD ALFAH57358.03
3-Chloro-N-(3-fluorobenzyl)-4-methoxyaniline, 97%
Purity: 97%
CAS No.: 1019595-79-9
Molecular Formula: C14H13ClFNO
Formula Weight: 265.71
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR