Tisk…

N-(2-Fluorobenzyl)-4-iodoaniline, 97%

H57929.MDEA 763 CZK
ALFAH57929.MD ALFAH57929.03
N-(2-Fluorobenzyl)-4-iodoaniline, 97%
Purity: 97%
CAS No.: 934364-17-7
Molecular Formula: C13H11FIN
Formula Weight: 327.14
MDL No.: MFCD11179979
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR