Tisk…

2,5-Difluoro-N-(4-fluorobenzyl)aniline, 97%

H57209.03EA 9770 CZK
ALFAH57209.03 ALFAH57209.MD
2,5-Difluoro-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019585-04-6
Molecular Formula: C13H10F3N
Formula Weight: 237.22
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR