Tisk…

4-Nitro-2,3-lutidine-N-oxide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL559849-25GEA 8130 CZK
SIAL559849-25G
4-Nitro-2,3-lutidine-N-oxide, Sigma-Aldrich®
4-Nitro-2,3-lutidine-N-oxide
Vzorec: C₇H₈N₂O₂
MW: 168,15 g/mol
MDL Number: MFCD00065172
CAS číslo: 37699-43-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR