Tisk…

4-Amino-2-chloro-6,7-dimethoxyquinazoline, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL518654-25GEA 9400 CZK
SIAL518654-25G
4-Amino-2-chloro-6,7-dimethoxyquinazoline, Sigma-Aldrich®
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Vzorec: C₁₀H₁₀ClN₃O₂
MW: 239,66 g/mol
MDL Number: MFCD00051734
CAS číslo: 23680-84-4
EINECS: 245-821-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR