Tisk…
Di-n-octyl phthalate, Supelco®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
48560-UEA 0 CZK
48560-U
Di-n-octyl phthalate, Supelco®
Di-n-octyl phthalate
Vzorec: C₂₄H₃₈O₄
MW: 390,56 g/mol
MDL Number: MFCD00015292
CAS číslo: 117-84-0
EINECS: 204-214-7