N,N'-Diisopropylethylenediamine, Sigma-Aldrich®
Dodavatel: Merck
SIAL298964-25GEA
12520
CZK
SIAL298964-25G
SIAL298964-5G
N,N'-Diisopropylethylenediamine, Sigma-Aldrich®
N,N'-Diisopropylethylenediamine
Vzorec:
C₈H₂₀N₂ MW: 144,26 g/mol |
MDL Number:
MFCD00008864 CAS číslo: 4013-94-9 EINECS: 223-666-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...