Tisk…

Dimethyl-4-nitrophthalate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL367486-5GEA 0 CZK
SIAL367486-5G
Dimethyl-4-nitrophthalate, Sigma-Aldrich®
Dimethyl-4-nitrophthalate
Vzorec: C₁₀H₉NO₆
MW: 239,19 g/mol
MDL Number: MFCD00017191
CAS číslo: 610-22-0
EINECS: 210-212-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR