5-Bromo-4,6-dichloropyrimidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL684635-1GEA
5790
CZK
SIAL684635-1G
5-Bromo-4,6-dichloropyrimidine, Sigma-Aldrich®
5-Bromo-4,6-dichloropyrimidine
Vzorec:
C₄HBrCl₂N₂ MW: 227,88 g/mol |
MDL Number:
MFCD00047379 CAS číslo: 68797-61-5 EINECS: 272-305-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...