Tisk…

Isopropyldiphenylphosphine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL336920-2GEA 3380 CZK
SIAL336920-2G
Isopropyldiphenylphosphine, Sigma-Aldrich®
Isopropyldiphenylphosphine
Vzorec: C₁₅H₁₇P
MW: 228,27 g/mol
MDL Number: MFCD00015028
CAS číslo: 6372-40-3
EINECS: 228-902-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR