Tisk…

křemík, powder, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL267414-25GEA 0 CZK
SIAL267414-25G
křemík, powder, Sigma-Aldrich®
křemík
Vzorec: Si
MW: 28,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS číslo: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR