Tisk…

5-Hydroxy-o-toluic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL696366-1GEA 2610 CZK
SIAL696366-1G
5-Hydroxy-o-toluic acid, Sigma-Aldrich®
5-Hydroxy-o-toluic acid
Vzorec: C₈H₈O₃
MW: 152,15 g/mol
MDL Number: MFCD09833184
CAS číslo: 578-22-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR