Tisk…

Benzyl chloromethyl ether

Dodavatel: ABCR GMBH

ABCRAB135117.0025EA 0 CZK
ABCRAB135117.0025
Benzyl chloromethyl ether
Benzyl chloromethyl ether
Vzorec: C₈H₉ClO
MW: 156,61 g/mol
MDL Number: MFCD00000886
CAS číslo: 3587-60-8
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR