Tisk…

4-(2-Methoxyethyl)phenol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL534595-25GEA 5630 CZK
SIAL534595-25G
4-(2-Methoxyethyl)phenol, Sigma-Aldrich®
4-(2-Methoxyethyl)phenol
Vzorec: C₉H₁₂O₂
MW: 152,19 g/mol
MDL Number: MFCD00017537
CAS číslo: 56718-71-9
EINECS: 260-354-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR