Tisk…

trans-1-Bromo-4-(4-ethylcyclohexyl)benzene, 97%

H55969.06EA 6100 CZK
ALFAH55969.06 ALFAH55969.03
trans-1-Bromo-4-(4-ethylcyclohexyl)benzene, 97%
Purity: 97%
CAS No.: 91538-82-8
Molecular Formula: C14H19Br
Formula Weight: 267.21
Melting Point: 30-32°
MDL No.: MFCD08275628
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR