Homophthalic anhydride
MOLEM83210533EA
0
CZK
MOLEM83210533
Homophthalic anhydride
Homophthalic anhydride
Vzorec:
C₉H₆O₃ MW: 162,14 g/mol |
MDL Number:
MFCD00006894 CAS číslo: 703-59-3 EINECS: 211-873-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...