DL-Glutamic acid
Dodavatel: EHRENSTORFER GMBH
EHERC14034400EA
1300
CZK
EHERC14034400
DL-Glutamic acid
DL-Glutamic acid
Vzorec:
C₅H₉NO₄ MW: 147,13 g/mol |
MDL Number:
MFCD00063113 CAS číslo: 617-65-2 EINECS: 210-522-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...