Tisk…

Indium(III) hydroxide, Sigma-Aldrich®

Dodavatel: Merck

SIAL318477-10GEA 0 CZK
SIAL318477-10G
Indium(III) hydroxide, Sigma-Aldrich®
Indium(III) hydroxide
Vzorec: H₃InO₃
MW: 165,84
MDL Number: MFCD00011056
CAS číslo: 20661-21-6
EINECS: 243-947-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR