Tisk…

Ethyl-3,5-bis(trifluoromethyl)phenylacetate ≥98%

Dodavatel: Thermo Scientific
Varování

H25820.06EA 4530 CZK
H25820.06 H25820.14 H25820.03
Ethyl-3,5-bis(trifluoromethyl)phenylacetate ≥98%
Ethyl-3,5-bis(trifluoromethyl)phenylacetate
Vzorec: C₁₂H₁₀F₆O₂
MW: 300,20
Storage Temperature: Ambient
MDL Number: MFCD08689894
CAS číslo: 144632-97-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR