Tisk…

Sodium bis(trimethylsilyl)amide

Dodavatel: ABCR GMBH

ABCRAB134636.0100EA 0 CZK
ABCRAB134636.0100
Sodium bis(trimethylsilyl)amide
Sodium bis(trimethylsilyl)amide
Vzorec: C₆H₁₈NNaSi₂
MW: 183,38 g/mol
Hustota: 1,065 g/cm³ (20 °C)
MDL Number: MFCD00009835
CAS číslo: 1070-89-9
EINECS: 213-983-8
UN: 3096
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR