Silver tetrafluoroborate ≥99%
Dodavatel: ABCR GMBH
|
Nebezpečí
|
ABCRAB121978.0005EA
0
CZK
ABCRAB121978.0005
Silver tetrafluoroborate ≥99%
Silver tetrafluoroborate
Vzorec:
AgBF₄ MW: 194,67 g/mol Bod tání: 200 °C (rozkl.) |
MDL Number:
MFCD00003408 CAS číslo: 14104-20-2 EINECS: 237-956-5 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...