Tisk…

Silver trifluoroacetate ≥98%

Dodavatel: ABCR GMBH
Varování

Synonyms: Silver(I) 2,2,2-trifluoroacetateSilver(I) trifluoroacetate

ABCRAB121980.0025EA 0 CZK
ABCRAB121980.0025
Silver trifluoroacetate ≥98%
Silver trifluoroacetate
Vzorec: C₂AgF₃O₂
MW: 220,88 g/mol
MDL Number: MFCD00013199
CAS číslo: 2966-50-9
EINECS: 221-004-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR