Order Entry
Lead standard solution, 10000 mg/L Pb in 5% nitric acid standard for ICP
Lead standard solution, 10000 mg/L Pb in 5% nitric acid standard for ICP
Supplier:  CPACHEM
Certificates
Lead standard solution, 10000 mg/L Pb in 5% nitric acid standard for ICP
Supplier:  CPACHEM
Product Information
  • Catalog #
    Supplier #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • ACSDC141.5NP.L5
    C141.5NP.L5
    Lead
    Pb
    10000 mg/L
    5% HNO₃
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL
    Supplier Article Number: C141.5NP.L5
    Local Article Number: ACSDC141.5NP.L5
  • ACSDC141.5NP.L1
    C141.5NP.L1
    Lead
    Pb
    10000 mg/L
    5% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL
    Supplier Article Number: C141.5NP.L1
    Local Article Number: ACSDC141.5NP.L1
  • ACSDC141.5NP.L25
    C141.5NP.L25
    Lead
    Pb
    10000 mg/L
    5% HNO₃
    Plastic bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL
    Supplier Article Number: C141.5NP.L25
    Local Article Number: ACSDC141.5NP.L25