Tisk…

Chloromethyl chloroformate

Dodavatel: ABCR GMBH

ABCRAB139652.0025EA 0 CZK
ABCRAB139652.0025
Chloromethyl chloroformate
Chloromethyl chloroformate
Vzorec: C₂H₂Cl₂O₂
MW: 128,94 g/mol
MDL Number: MFCD00077688
CAS číslo: 22128-62-7
EINECS: 244-793-3
UN: 2745
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR