Tisk…

Ethanol 30,0% (v/v)

Nebezpečí

VWRS83909.550
Ethanol 30,0% (v/v)
Ethanol
Vzorec: C₂H₅OH
MW: 46,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS číslo: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,III
Merck Index: 13,03795

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR