Tisk…
Industrial gloves, G12
Dodavatel: Marigold (Ansell)
112-2831EA 815 CZK
112-2831
Industrial gloves, G12
Rukavice
Durable, unsupported natural rubber gloves with flock lining.
  • For use in low hazard areas
  • Beaded cuff for tear resistance and easy pull on
  • Good abrasion resistance
The gloves can be used for janitorial applications, food handling & processing, manufacturing and maintenance, and in laboratories.
Thickness: 0,85 mm (double wall)
EN 388 (2010), EN 374, Cat. III; AQL \< 0,65
Food contact approved
Certifikace: EN 388 (2010), EN 374, Cat. III; AQL < 0,65
Food contact approved