Tisk…
Heat resistant gloves, IHR 040
Dodavatel: Honeywell Safety Products
112-2942EA 0 CZK
112-2942
Heat resistant gloves, IHR 040
This heat resistant glove is made from a two layer Kevlar® lining, with glass fibre in-between, and finished with a rubber/silicone coating.
  • Alternating Kevlar® and glass fibre offers excellent insulation with respect to the contact temperature
  • Good protection against splashes of molten metal
  • Aluminised glass fibre cuff protects a large part of the forearm
  • Ambidextrous
  • Heat resistant up to 900 °C
The glove withstands 900 °C for up to 20 seconds and is suitable for quickly grasping and moving hot parts, for example in metallurgy and glass industries, or in foundries.
Certifikace: EN 388 (2040), EN 407 (44XX34); Cat. II
Upozornení: *One size fits all