Tisk…
Welding sleeve protectors, SW20, leather
Dodavatel: Portwest
Portwest®
113-7630EA 546 CZK
113-7630
Welding sleeve protectors, SW20, leather
Pracovní oděvy
These welding sleeves made of split cowhide leather with Kevlar® stitching are designed to protect the forearm and elbow in the toughest environment.
  • Elasticised ends ensures maximum protection and a secure fit
  • Ideal for combination with the welding apron SW10
Certifikace: EN ISO 11611 (2 A1)