Ethanol 50% (v/v)
|
Nebezpečí
|
VWRS5988.9025
Ethanol 50% (v/v)
Ethanol
Vzorec:
C₂H₅OH MW: 46,07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS číslo: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...