Tisk…
Rain jacket, Portwest Texo, TX30
Dodavatel: Portwest
113-7644EA 2400 CZK
PRTWTX30GRRL PRTWTX30GRRM
Rain jacket, Portwest Texo, TX30
Waterproof and durable jacket with hood, made of polyester with a PVC coating and polyester lining.
  • Waterproof taped seams
  • Velcro® cuffs
  • Detachable ID pocket, mobile phone pocket, pen pockets and multiple other pockets
Weight: 250 g/m²; lining: 170 g/m²
EN 343 (3-1)
Certifikace: EN 343 (3-1)