Tisk…
Flame-retardant shirts, long sleeved, Teramo 2690
Dodavatel: Sioen
113-7055EA 1440 CZK
SIOE2690AP1BS2XL SIOE2690AP1BS3XL SIOE2690AP1BSL SIOE2690AP1BSM SIOE2690AP1BS1XL SIOE2690AP1BSS
Flame-retardant shirts, long sleeved, Teramo 2690
Soft shirt made of 60% Protex® and 40% Viloft® (single jersey). The inherently flame-retardant fabric has good moisture regulating properties and features an anti-bacterial treatment.
  • Lightweight
  • Round neck, long sleeves
  • Elasticated wristband
Weight: 180 g/m²
Certifikace: EN ISO 11612 (A1 B1 C1 F1), EN 14116 (index 3)