Tisk…
Cleanroom gloves, nitrile, Showa C9905PF
Dodavatel: Showa
N-Dex®
112-3317EA 45190 CZK
BESTC9905PFS BESTC9905PFXS BESTC9905PFL BESTC9905PFM BESTC9905PFXL
Cleanroom gloves, nitrile, Showa C9905PF
These ambidextrous gloves protect the hand from a wide array of chemical hazards and deliver low ESD and particulate counts, protecting high-end semi-conductors and perishable food-stuffs alike.
  • Non sterile, powder-free, smooth finish
  • Rolled cuff prevents dirt from entering the glove
  • Surface provides good grip and repels wet substances
  • Antistatic properties: Suitable for handling of electronic devices
  • Great freedom of movement for the wrist
The gloves are ideal for cleanroom, lab and food processing environments where exacting standards of industrial hygiene are required.
Thickness: 0,12 mm
Colour: White
Cat. I
Certifikace: EN ISO 374-1 Type C (K), EN ISO 374-5 VIRUS; Cat. III
Balení: 100 gloves per bag.