4-Isobutyl-3-nitrophenylboronic acid
Dodavatel: Apollo Scientific
APOSOR5535-1GEA
7710
CZK
APOSOR5535-1G
4-Isobutyl-3-nitrophenylboronic acid
4-Isobutyl-3-nitrophenylboronic acid
Vzorec:
C₁₀H₁₄BNO₄ |
MDL Number:
MFCD04115653 CAS číslo: 850568-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...