Tisk…

Piperidine-[D11] (98% D), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL448141-1GEA 10780 CZK
SIAL448141-1G
Piperidine-[D11] (98% D), Sigma-Aldrich®
Piperidine-[D11]
Vzorec: C₅D₁₁N
CAS číslo: 143317-90-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR