Scandium(III) perchlorate 40% vodný roztok, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL444146-5GEA
6140
CZK
SIAL444146-5G
Scandium(III) perchlorate 40% vodný roztok, Sigma-Aldrich®
Scandium(III) perchlorate
Vzorec:
Cl₃O₁₂Sc MW: 451,42 g/mol (343,30 anhy) |
MDL Number:
MFCD00243289 CAS číslo: 14066-05-8 EINECS: 237-914-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...