Tetraethylammonium nitrate 35% (w/w) vodný roztok
|
Nebezpečí
|
41791.ADEA
975
CZK
41791.AD
41791.AK
Tetraethylammonium nitrate 35% (w/w) vodný roztok
Tetraethylammonium nitrate
Vzorec:
C₈H₂₀N₂O₃ MW: 192,26 g/mol Bod varu: >100 °C Hustota: 1,029 Storage Temperature: Ambient |
MDL Number:
MFCD00041978 CAS číslo: 1941-26-0 EINECS: 217-725-5 UN: 3139 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...