Tisk…

Tetrahydropyran-2-methanol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIALT14605-5GEA 1330 CZK
SIALT14605-5G
Tetrahydropyran-2-methanol, Sigma-Aldrich®
Tetrahydropyran-2-methanol
Vzorec: C₆H₁₂O₂
MW: 116,16 g/mol
MDL Number: MFCD00006624
CAS číslo: 100-72-1
EINECS: 202-882-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR