Tisk…

Cholesterol oleyl carbonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL151157-25GEA 4090 CZK
SIAL151157-25G
Cholesterol oleyl carbonate, Sigma-Aldrich®
Cholesterol oleyl carbonate
Vzorec: C₄₆H₈₀O₃
MDL Number: MFCD00003634
CAS číslo: 17110-51-9
EINECS: 241-179-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR