Tisk…

Tritylium hexachloroantimonate ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

B25035.22EA 8650 CZK
B25035.22 B25035.06 B25035.14
Tritylium hexachloroantimonate ≥99%
Tritylium hexachloroantimonate
Vzorec: C₁₉H₁₅Cl₆Sb
MW: 577,80
Bod tání: ∼ 220 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00013126
CAS číslo: 1586-91-0
EINECS: 216-446-6
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR