Tisk…

Acetic anhydride-[¹³C₄] (99% ¹³C ), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL487821-250MGEA 0 CZK
SIAL487821-250MG
Acetic anhydride-[¹³C₄] (99% ¹³C ), Sigma-Aldrich®
Acetic anhydride-[¹³C₄]
Vzorec: C₄H₆O₃
MDL Number: MFCD00083864
CAS číslo: 114510-14-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR