Tisk…
Fluorophosphoric acid ≥70%, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL324744-100GEA 0 CZK
SIAL324744-100G
Fluorophosphoric acid ≥70%, Sigma-Aldrich®
Fluorophosphoric acid
Vzorec: H₂FO₃P
MW: 99,99 g/mol
MDL Number: MFCD00011334
CAS číslo: 13537-32-1
UN: 1776
ADR: 8,II