Tisk…

3-(2,3-Dimethylphenylcarbamoyl)benzeneboronic acid, 96%

H52762.MDEA 4150 CZK
ALFAH52762.MD ALFAH52762.03
3-(2,3-Dimethylphenylcarbamoyl)benzeneboronic acid, 96%
Purity: 96%
CAS No.: 957060-99-0
Molecular Formula: C15H16BNO3
Formula Weight: 269.11
MDL No.: MFCD09027262
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR