Tisk…

3-(2,4-Dimethylphenylcarbamoyl)benzeneboronic acid, 98%

H53293.MDEA 830 CZK
ALFAH53293.MD ALFAH53293.03
3-(2,4-Dimethylphenylcarbamoyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 957060-98-9
Molecular Formula: C15H16BNO3
Formula Weight: 269.11
MDL No.: MFCD09027261
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR