Tisk…

3-Ethoxycarbonyl-5-nitrobenzeneboronic acid, 97%

H52873.03EA 897 CZK
ALFAH52873.03 ALFAH52873.06
3-Ethoxycarbonyl-5-nitrobenzeneboronic acid, 97%
Purity: 97%
CAS No.: 850568-37-5
Molecular Formula: C9H10BNO6
Formula Weight: 238.99
Melting Point: 158-163°
MDL No.: MFCD03788422
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR