Tisk…
4-Chloro-3-(2,3-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
H54631.03EA 8140 CZK
ALFAH54631.03 ALFAH54631.MD
4-Chloro-3-(2,3-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD20265303