Tisk…
4-Chloro-3-(2,4-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
H54912.03EA 4890 CZK
ALFAH54912.03 ALFAH54912.MD
4-Chloro-3-(2,4-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD20265372