Tisk…

2-Chloro-5-(2,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%

H54999.03EA 7000 CZK
ALFAH54999.03 ALFAH54999.MD
2-Chloro-5-(2,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR